Agarose, NuSieve® Genetic Technology Grade™ (GTG)
Leverandør: LONZA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50080 50081 50084
733-1543EA 14020 DKK
733-1543 733-1544 733-1545
Agarose, NuSieve® Genetic Technology Grade™ (GTG)
Elektroforese geler Agarose og Akrylamid
NuSieve® Genetic Technology Grade™ (GTG) Agarose is a low melting temperature (65 °C) agarose that finely resolves DNA fragments, PCR and RT-PCR products ranging from 10 to 1000 bp.
  • High gel strength makes gels easy to handle, enhancing the convenience of gel processing and blotting
  • Performance tested to ensure fine resolution of DNA fragments up to 1000 bp
  • DNAse- and RNAse-free
Cloning procedures can be performed directly in remelted NuSieve® GTG™ Agarose, eliminating costly and time-consuming DNA extraction steps. This agarose is also compatibleMore Product Information