Agarose, SeaKem® LE
Leverandør: LONZA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50000 50004 50005 50001 50002
733-1535EA 4170 DKK
733-1535 733-0829 733-1538 733-1536 733-1537
Agarose, SeaKem® LE
Elektroforese geler Agarose og Akrylamid
SeaKem® LE is a high gel strength, low EEO, standard gelling temperature agarose perfect for electrophoresis and other molecular biology applications.
  • Quality tested to certify performance
  • High gel strength
  • Low EEO
  • Standard melting temperature
  • DNase- and RNase-free
SeaKem® LE agarose is ideal for routine analysis of nucleic acids by gel electrophoresis and blotting. Each SeaKem LE gel sharply resolves DNA and provides consistent resolution from lot-to-lot. This molecular biology grade agarose has no detectable DNase or RNase activity and forms strongMore Product Information