Connectors and ferrules, Gerstel GRAPHPACK® 3D/2
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20272 20270 20271 20273 20274
REST20272EA 0 DKK
REST20272 REST20270 REST20271 REST20273 REST20274
Connectors and ferrules, Gerstel GRAPHPACK® 3D/2
Fittings og stik
GRAPHPACK® technology provides a complete system that quickly and reliably makes a leak-tight, low-dead-volume connection. The central component is a metal-jacketed graphite ferrule - the ideal seal for GC applications.