Lab air quality sensor, Halo Sense
Leverandør: ERLAB CAPTAIRE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AS016010021 AS036020021 AS046030022
ERLAAS016010021EA 8030 DKK
ERLAAS016010021 ERLAAS036020021 ERLAAS046030022
Lab air quality sensor, Halo Sense
Overvågningssystem Miljøovervågningssystemer
Halo Sense has been designed to continuously monitor the lab air quality for a large spectrum of fumes and vapours. Halo Sense comes in 3 versions: Volatile Organic Compounds (VOC), acid and formaldehyde.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://dk.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=20525804
  • Simple communication by LED pulsation system
  • Power supplies in compliance with various international standards
  • RJ45 cable provided
  • Default settings: audible alarm disabled, visual alarm activated (both alarms can be disabled)
  • To be installed: on the ceiling, on the wall, on the bench, or under
When Halo senses fumes or vapours, a soft band of light pulses to let the user know that the air is polluted.
The monitoring gives the user the control to monitor the device and it will send alerts to the preferred device. If the laboratory air is polluted, it allows the user to take action before pollution levels become a hazard.
Filtration fault detection: Detection sensors for solvents, acids or formaldehyde.
Advarsel: There is a very wide range of Erlab products. To ensure you receive the correct product for your specific application and setting please complete the contact form as per the url above. Your local Avantor sales representative will then send you an eValiQuest form to get details of your application and requirements. Once we have received this back from you, we will send you a quotation based on your precise requirements ensuring all handling and safety requirements are met. 