Cut resistant gloves, HyFlex® 11-531 / 11-537 / 11-539
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-537/9 11-537/10 11-537/11 11-537/6 11-531/6 11-531/7 11-531/8 11-531/9 11-531/10 11-531/11 11-537/7 11-537/8
ANSE11-537/9EA 1540 DKK
ANSE11-537/9 ANSE11-537/10 ANSE11-537/11 ANSE11-537/6 ANSE11-531/6 ANSE11-531/7 ANSE11-531/8 ANSE11-531/9 ANSE11-531/10 ANSE11-531/11 ANSE11-537/7 ANSE11-537/8
Cut resistant gloves, HyFlex® 11-531 / 11-537 / 11-539
Handsker
These seamless plaited liners with technology yarn feature a partial (palm and fingertips or ¾ dipped) or a full nitrile foam coating.
  • INTERCEPT™ cut resistance yarn has been designed to exceed industry standards
  • Best-in-class cut protection and increased comfort with dexterity
  • ZONZ™ Comfort Fit technology for enhanced fit, higher dexterity and reduced hand fatigue
  • FORTIX® formulation provides enhanced abrasion resistance and grip, which extends gloves’ working life and improves comfort in abrasive applications
  • Silicone-free
  • Dirt masking colours for longer wear times
Ideal for the assembly of small sharp parts, handling small sharp metal parts, light duty stamping and maintenance.
Certificeringer: EN 388:2016 (4X42B); Cat. II