General purpose gloves, AlphaTec® 37-200 / 37-210
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37-200/11 37-200/12 37-200/9 37-200/7 37-210/8
ANSE37-200/11EA 0 DKK
ANSE37-200/11 ANSE37-200/12 ANSE37-200/9 ANSE37-200/7 ANSE37-210/8
General purpose gloves, AlphaTec® 37-200 / 37-210
Handsker
These lightweight, reusable nitrile gloves are designed specifically for food processing and handling.
  • Ansell UltraGrip™ technology: Thanks to the raised diamond-pattern surfacing it offers good grip in wet and oily conditions
  • Outstanding flexibility and dexterity, even at low temperature
  • Can be used on their own or as outer gloves over thermal liners or cut resistant gloves
  • Latex-free formulation prevents risk of Type 1 allergies
  • Liquid-proof
Ideal for use in poultry processing, fruit and vegetable processing, dairy processing and for preparing meals.
Certificeringer: EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5 VIRUS, EN 388:2016 (2001X); Cat. III
Food contact approved.