Industrial gloves, oil repellent, ActivArmr® Hycron® 27-905
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
27-905/11 27-905/9 27-905/8 27-905/10
ANSE27-905/11EA 592 DKK
ANSE27-905/11 ANSE27-905/9 ANSE27-905/8 ANSE27-905/10
Industrial gloves, oil repellent, ActivArmr® Hycron® 27-905
Handsker
These gloves with improved nitrile coating formulation and polycotton lining are more durable than ever providing superior resistance to abrasive and sharp materials.
  • Fortix® Technology: Thin, resilient and breathable nitrile foam coating that extends the glove's working life and improves the comfort in abrasive applications
  • Ansell Grip Technology™: Excellent grip in dry, wet and oily environments
  • Ripel™ Technology: Outstanding barrier to grease and oil; glove does not degrade like leather or cotton
Ideal for garbage collection and sorting, handing abrasive and low cut materials, handling dry, wet and oily large objects, maintenance and primary assembly.
Certificeringer: EN 388:2016 (4221C); Cat. II.