Septum nuts
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21309 20631
REST21309EA 538 DKK
REST21309 REST20631
Septum nuts
Fittings og stik
These septum nuts are made of high-quality stainless steel and for use with Agilent 5890/6850/6890/7890 split or splitless injectors.
  • Needle guide allows easy penetration and prevents septum coring
  • Manual injection septum nut allows use of 26-gauge needles for on-column injections