Soap film flow meters
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20135 20136
REST20135EA 635 DKK
REST20135 REST20136
Soap film flow meters
Flow-metre
Both flow meters are suitable for applications with soap.
Bestillingsoplysninger: Include a reservoir bulb, 600 mm of 1/4" ID tubing, adapter tubes for 1/8" tubing and 0,53 mm ID capillary columns, and Velcro fasteners.