Preparative HPLC columns, Ultra Silica
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9100517 9100518 9100519 9100557 9100558 9100559 9100567 9100568 9100569 9100577 9100578 9100579
REST9100517EA 0 DKK
REST9100517 REST9100518 REST9100519 REST9100557 REST9100558 REST9100559 REST9100567 REST9100568 REST9100569 REST9100577 REST9100578 REST9100579
Preparative HPLC columns, Ultra Silica
Kromatografikolonner
Base-deactivated spherical silica is useful for normal-phase or HILIC separations.