VWR® Traceable®, Remote Alarm Thermo-Hygrometer
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-2486EA 1040 DKK
620-2486 620-2487
VWR® Traceable®, Remote Alarm Thermo-Hygrometer
Hygrometre Digitale termo-hygrometre
These thermo-hygrometers are ideal for routine measurements, monitoring 'round the clock', all quality control needs, and critical experimental requirements. Perfect for use in hoods, classrooms, stockrooms, environmental chambers and chemical storage areas.
  • Triple-display shows min., max. and current humidity/temperature readings
  • Min./max. temperature and humidity memories
  • High/low alarm feature may be set by user in 1% RH or 1° increments
  • F/°C switchable
  • Remote humidity/temperature sensor on a 2,1 metre cable
* Temperature
** Humidity
***Temperature probe
Certificeringer: Individually calibrated ISO 17025 certified. Individually certified—traceable to NIST.
Leveringsinformation: Supplied with Traceable® certificate, battery, external sensor, flip-open stand and wall mount.