Intermediate-polarity deactivated guard/retention gap columns/transfer lines (fused silica)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10041 10042 10043 10044 10045 10046 10047 10048 10049 10032 10043-600 10044-600 10045-600 10102 10046-600 10048-600 10049-600 10097 10098 10099 10100 10101 10012 10013 10022 10023 10033
REST10041EA 1070 DKK
REST10041 REST10042 REST10043 REST10044 REST10045 REST10046 REST10047 REST10048 REST10049 REST10032 REST10043-600 REST10044-600 REST10045-600 REST10102 REST10046-600 REST10048-600 REST10049-600 REST10097 REST10098 REST10099 REST10100 REST10101 REST10012 REST10013 REST10022 REST10023 REST10033
Intermediate-polarity deactivated guard/retention gap columns/transfer lines (fused silica)
Kromatografikolonner
Intermediate-polarity deactivated guard/retention gap columns are useful for a wide range of applications and are proven to be compatible with most common solvents.
  • Tested with a comprehensive test mix to ensure high inertness
  • Useful for a wide range of applications
  • Use with most common solvents
  • Maximum temperature: 325 °C
  • 30- and 60-meter lengths are banded in 5-meter sections