Gas sampling bags, ALTEF®
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22963 22962 22959 22960 26328 26329 26330 26331 22961
REST22963EA 3610 DKK
REST22963 REST22962 REST22959 REST22960 REST26328 REST26329 REST26330 REST26331 REST22961
Gas sampling bags, ALTEF®
Poser Gasposer
Gas sampling bags are whole air sampling devices useful for monitoring part-per million (ppm) levels of most volatile organic compounds (VOCs). Unlike Tedlar® bags, ALTEF bags do not produce background levels of dimethylacetamide (DMAC) or phenol. It features single eyelet and PP combo valve with hose connection to fit 4,7 mm ID tubing and syringe port with replaceable septum.
  • Durable polyvinylidene fluoride (PVDF) film is resistant to abrasion and chemicals, including most acids and organic compounds
  • Excellent alternative to Tedlar® bags for collection of most VOCs
  • Very low VOC and sulfur background compared to Tedlar® bags
  • Not recommended for ketones, acetates, hydrogen sulfide or permanent gases
  • Contain no additives, fillers, or pigments
  • 82 °C (180 °F) maximum operating temperature
Note: Use only Point Style 5 syringes.