Filters for extraction cells, ASE® 200 systems
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26119 26118 26190
REST26119EA 500 DKK
REST26119 REST26118 REST26190
Filters for extraction cells, ASE® 200 systems
Filtre Glasfiberfiltre
These filter are made of cellulose or glass fibre. Ideal for ASE® 200 extraction cells.