QuEChERS extraction salt packets Q-sep™
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26236 26237 26238 26239 23992
REST26236EA 0 DKK
REST26236 REST26237 REST26238 REST26239 REST23992
QuEChERS extraction salt packets Q-sep™
Kromatografikolonner
Q-sep™ QuEChERS provides fast, simple sample preparation for multi residue pesticide analysis.
  • Preweighed, ultra-pure sorbents
  • Convenient and easy to use
  • Salt packets eliminate the need for a second empty tube to transfer salts
Support original unbuffered, AOAC (2007.01) and European (EN 15662) QuEChERS methods.