Anti-KRT8 & KRT18 Mouse Monoclonal Antibody [clone: B22.1 B23.1]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNUB1221-500 BNUB1221-100
BNUB1221-500EA 3810 DKK
BNUB1221-500 BNUB1221-100
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody [clone: B22.1 B23.1]
Antistoffer
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This MAb cocktail recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 1More Product Information