Anti-CD74 Mouse Monoclonal Antibody [clone: LN-2 CLIP/813]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNUB1207-500 BNUB1207-100
BNUB1207-500EA 3810 DKK
BNUB1207-500 BNUB1207-100
Anti-CD74 Mouse Monoclonal Antibody [clone: LN-2 CLIP/813]
Antistoffer
This antibody recognizes a protein of ~35 kDa, identified as CD74. It is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74 has been shown to be useful in differentiating atypical fibroxanthoma (-) from malignant fibrous histiocytoma ( ).

  • Purified

Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: LN-2 CLIP/813
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR