Anti-EGFR Mouse Monoclonal Antibody (CF594) [clone: 31G7]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC941194-500 BNC941194-100
BNC941194-500EA 4180 DKK
BNC941194-500 BNC941194-100
Anti-EGFR Mouse Monoclonal Antibody (CF594) [clone: 31G7]
Antistoffer
This MAb recognizes a protein of 170 kDa, identified as EGFR. EGFR is type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 31G7
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR