Anti-MYH11 Mouse Monoclonal Antibody (Biotin) [clone: MYH11/923 SMMS-1]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNCB1136-500 BNCB1136-100
BNCB1136-500EA 4180 DKK
BNCB1136-500 BNCB1136-100
Anti-MYH11 Mouse Monoclonal Antibody (Biotin) [clone: MYH11/923 SMMS-1]
Antistoffer
Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors.

Type: Primary
Antigen: MYH11
Clonality: Monoclonal
Clone: MYH11/923 SMMS-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR