Anti-CR2 Mouse Monoclonal Antibody (CF488A) [clone: FR5A10]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC881087-500 BNC881087-100
BNC881087-500EA 4180 DKK
BNC881087-500 BNC881087-100
Anti-CR2 Mouse Monoclonal Antibody (CF488A) [clone: FR5A10]
Antistoffer
Recognizes a protein of 140 kDa, which is identified as the complement receptor 2 (CR2)/CD21. Its epitope is located in 5-8 short consensus repeats (SCRs). This MAb is highly specific to CR2 and shows no cross-reaction with CR1. This protein is expressed strongly on mature B cells, follicular dendritic cells and weakly on immature thymocytes and T lymphocytes. In B-cell ontogeny, CD21 appears after the pre-B-stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD21 expression is also gradually lost after stimulation of B cellsMore Product Information