Anti-CR2 Mouse Monoclonal Antibody (Purified) [clone: FR5A10]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNUB1087-500 BNUB1087-100
BNUB1087-500EA 3810 DKK
BNUB1087-500 BNUB1087-100
Anti-CR2 Mouse Monoclonal Antibody (Purified) [clone: FR5A10]
Antistoffer
Recognizes a protein of 140 kDa, which is identified as the complement receptor 2 (CR2)/CD21. Its epitope is located in 5-8 short consensus repeats (SCRs). This MAb is highly specific to CR2 and shows no cross-reaction with CR1. This protein is expressed strongly on mature B cells, follicular dendritic cells and weakly on immature thymocytes and T lymphocytes. In B-cell ontogeny, CD21 appears after the pre-B-stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD21 expression is also gradually lost after stimulation of B cells in vitro. CD21 functions as receptor for C3d, C3dg and iC3b Complement components, for EBV and for IFNalpha. CD21 binds to CD23 and associates with CD19, CD81 and Leu13 to form a large signal-transduction complex involved in B cell activation. MAb FR5A10 can be used for EBV receptor studies, interactions between B and T cells especially through CD23, human complement receptor (CR2) studies and IFN-alpha receptor studies.

Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: FR5A10
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR