Anti-ITGA2B Mouse Monoclonal Antibody (CF647) [clone: ITGA2B/1036]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC471036-500 BNC471036-100
BNC471036-500EA 4180 DKK
BNC471036-500 BNC471036-100
Anti-ITGA2B Mouse Monoclonal Antibody (CF647) [clone: ITGA2B/1036]
Antistoffer
Reacts with a calcium-dependent complex of CD41/CD61, a dimer of 90 kDa and 140 kDa present on the membrane of normal platelets and megakaryocytes. CD41/CD61 is also known as platelet glycoprotein GPIIb/GPIIIa or integrin IIa/3. This complex is the receptor of fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation.

Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: ITGA2B/1036
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR