Anti-IFNA1 Mouse Monoclonal Antibody (Purified) [clone: 2.52]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNUB0998-500 BNUB0998-100
BNUB0998-500EA 3810 DKK
BNUB0998-500 BNUB0998-100
Anti-IFNA1 Mouse Monoclonal Antibody (Purified) [clone: 2.52]
Antistoffer
This MAb is specific for human interferon alpha 1 and does not cross react with human interferon alpha 2. Interferons are widely used therapeutic agents because of their anti-tumor and anti-viral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon- & Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is influenced by phosphorylation of Serine 535 and 539 in the IFNAR1.

Type: Primary
Antigen: IFNA1
Clonality: Monoclonal
Clone: 2.52
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR