Anti-SOX10 Mouse Monoclonal Antibody (CF594) [clone: SOX10/991]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC940991-500 BNC940991-100
BNC940991-500EA 4180 DKK
BNC940991-500 BNC940991-100
Anti-SOX10 Mouse Monoclonal Antibody (CF594) [clone: SOX10/991]
Antistoffer
Recognizes a protein of ~55 kDa, identified as SOX10. This MAb is highly specific and does not cross-react with other members of the SOX-family. SOX genes comprise a family of genes that are related to the mammalian sex-determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. SOX-10 is a sensitive marker of melanoma, including conventional, spindled, and desmoplastic subtypes. It is expressed by metastatic melanomas and nodal capsular nevus in sentinel lymph nodes, but not by otherMore Product Information