Anti-KAT3B/p300 Mouse Monoclonal Antibody [clone: RW128]
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NB100-507 NB100-507SS
NOVUNB100-507EA 4470 DKK
NOVUNB100-507 NOVUNB100-507SS
Anti-KAT3B/p300 Mouse Monoclonal Antibody [clone: RW128]
Antistoffer
Mouse Monoclonal KAT3B/p300 Antibody (RW128). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat, Primate.
Type: Primary
Antigen: KAT3B/p300
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human