Anti-PTGDS Rabbit Polyclonal Antibody
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NB110-59909 NB110-59909SS
NOVUNB110-59909EA 4410 DKK
NOVUNB110-59909 NOVUNB110-59909SS
Anti-PTGDS Rabbit Polyclonal Antibody
Antistoffer
Rabbit Polyclonal Prostaglandin D Synthase/Lipocalin Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Bovine, Equine, Feline, Primate, Porcine, Rabbit.
Type: Primary
Antigen: Prostaglandin D Synthase/Lipocalin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human