Anti-H1 Mouse Monoclonal Antibody (CF568) [clone: HH1/957]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC680957-500 BNC680957-100
BNC680957-500EA 4180 DKK
BNC680957-500 BNC680957-100
Anti-H1 Mouse Monoclonal Antibody (CF568) [clone: HH1/957]
Antistoffer
Eukaryotic histones are basic and water-soluble nuclear proteins that form hetero-octameric nucleosome particles by wrapping 146 base pairs of DNA in a left-handed super-helical turn sequentially to form chromosomal fiber. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form the octamer; formed of two H2A-H2B dimers and two H3-H4 dimers, forming two nearly symmetrical halves by tertiary structure. Over 80% of nucleosomes contain the linker Histone H1, derived from an intronless gene that interacts with linker DNA between nucleosomes and mediates compaction into higherMore Product Information