Anti-KAT3B/p300 Mouse Monoclonal Antibody [clone: RW109]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NB100-617 NB100-617SS
NOVUNB100-617EA 4140 DKK
NOVUNB100-617 NOVUNB100-617SS
Anti-KAT3B/p300 Mouse Monoclonal Antibody [clone: RW109]
Antistoffer
Mouse Monoclonal KAT3B/p300 Antibody (RW109). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat, Primate.

Type: Primary
Antigen: KAT3B/p300
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR