Anti-CD14 Mouse Monoclonal Antibody (CF594) [clone: MDA/929]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC940929-500 BNC940929-100
BNC940929-500EA 4180 DKK
BNC940929-500 BNC940929-100
Anti-CD14 Mouse Monoclonal Antibody (CF594) [clone: MDA/929]
Antistoffer
Recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MDA/929
Conjugation: CF594
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG1, kappa
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR