Anti-PDCD1 Mouse Monoclonal Antibody (CF680) [clone: PDCD1/922]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC800922-500 BNC800922-100
BNC800922-500EA 4180 DKK
BNC800922-500 BNC800922-100
Anti-PDCD1 Mouse Monoclonal Antibody (CF680) [clone: PDCD1/922]
Antistoffer
PDCD-1 (programmed cell death-1 protein), also designated CD279, is a type I transmembrane receptor and a member of the immunoglobin gene superfamily. It is expressed on activated T-cells, B-cells, and myeloid cells. Anti-PDCD-1 is a marker of angioimmunoblastic lymphoma and suggests a unique cell of origin for this neoplasm. Unlike CD10 and BCL6, PDCD-1 is expressed by few B-cells, so anti-PDCD-1 may be a more specific and useful diagnostic marker in angioimmunoblastic lymphoma. In addition, PDCD-1 expression provides evidence that angioimmunoblastic lymphoma is a neoplasm derived from germinal center-associated T-cells.

Type: Primary
Antigen: PDCD1
Clonality: Monoclonal
Clone: PDCD1/922
Conjugation: CF680
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR