Anti-MYC Mouse Monoclonal Antibody (Biotin) [clone: MYC909]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNCB0909-500 BNCB0909-100
BNCB0909-500EA 4180 DKK
BNCB0909-500 BNCB0909-100
Anti-MYC Mouse Monoclonal Antibody (Biotin) [clone: MYC909]
Antistoffer
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or over-expressed in a variety of tumors. Over-expression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease. Rearrangement of the MYC gene is found in 3% to 16% of diffuse large B-cell lymphoma (DLBCL's) and in nearly 100% of Burkitt lymphomas (BL). Identifying MYC statusMore Product Information