Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: MYC909]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNUB0909-500 BNUB0909-100
BNUB0909-500EA 3810 DKK
BNUB0909-500 BNUB0909-100
Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: MYC909]
Antistoffer
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or over-expressed in a variety of tumors. Over-expression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease. Rearrangement of the MYC gene is found in 3% to 16% of diffuse large B-cell lymphoma (DLBCL's) and in nearly 100% of Burkitt lymphomas (BL). Identifying MYC statusMore Product Information
Bestil nu

Specifikationer

Antigen symbol MYC
Environmentally Preferable
Antigen name c-Myc
Antistof type Primary
Clonality Monoclonal
Konjugation Unconjugated
Clone Clone MYC909
Reactivity Human
Vært Mouse
Gen ID 202453
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunFluorescens Yes
Koncentration 0,2 mg/ml
Storage buffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR