Anti-CASP9 Rabbit Polyclonal Antibody

Leverandør: DIAGNOSTIC BIOSYSTEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


RP098 RP098-05
DIABRP098EA 0 DKK
DIABRP098 DIABRP098-05
Anti-CASP9 Rabbit Polyclonal Antibody
Antistoffer
Type:
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR