Anti-alpha-Actinin (Sarcomeric) Mouse Monoclonal Antibody [clone: EA-53]

Leverandør: DIAGNOSTIC BIOSYSTEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


MOB227 MOB227-05
DIABMOB227EA 0 DKK
DIABMOB227 DIABMOB227-05
Anti-alpha-Actinin (Sarcomeric) Mouse Monoclonal Antibody [clone: EA-53]
Antistoffer
Type:
Antigen: alpha-Actinin (Sarcomeric)
Clonality: Monoclonal
Clone: EA-53
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR