Anti-Ubiquitin Mouse Monoclonal Antibody [clone: FPM1]

Leverandør: DIAGNOSTIC BIOSYSTEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


MOB225 MOB225-05
DIABMOB225EA 0 DKK
DIABMOB225 DIABMOB225-05
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: FPM1]
Antistoffer
Type:
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: FPM1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR