Anti-F13A1 Polyclonal Antibody

Leverandør: DIAGNOSTIC BIOSYSTEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


RP103 RP103-05
DIABRP103EA 0 DKK
DIABRP103 DIABRP103-05
Anti-F13A1 Polyclonal Antibody
Antistoffer
This antibody reacts with a 160 kDa (unreacted dimer) and 80 kDa (reduced monomer) protein. Factor XIII is a β-globulin found in plasma and is composed of two subunits, Factor XIII-A and Factor XIII-B. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is a dermal dendrocyte marker and shows variable reaction with these types of tumors. Factor XIII can be used for histiocytic phenotyping and has been reported to mark capillary hemangiomas and tumors of the central nervous system.

Type:
Antigen: F13A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR