Anti-COX2 Rabbit Polyclonal Antibody

Leverandør: DIAGNOSTIC BIOSYSTEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


RP111 RP111-05
DIABRP111EA 0 DKK
DIABRP111 DIABRP111-05
Anti-COX2 Rabbit Polyclonal Antibody
Antistoffer
This antibody reacts with a 70 kDa protein. COX-2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters and cytokines that induce its expression. COX-2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX-1 levels remained unchanged.

Type:
Antigen: COX2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR