Anti-BCL2 Mouse Monoclonal Antibody (CF640R) [clone: BCL2/796]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC400796-500 BNC400796-100
BNC400796-500EA 4180 DKK
BNC400796-500 BNC400796-100
Anti-BCL2 Mouse Monoclonal Antibody (CF640R) [clone: BCL2/796]
Antistoffer
This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicularMore Product Information
Bestil nu

Specifikationer

Antigen symbol BCL2
Antigen name bcl-2
Environmentally Preferable
Antigen synonymer Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
Antistof type Primary
Clonality Monoclonal
Konjugation CF® 640R
Clone Clone BCL2/796
Reactivity Human
Vært Mouse
Gen ID 150749
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunFluorescens Yes
Koncentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR