Anti-PTPRC Mouse Monoclonal Antibody (CF568) [clone: 2B11 PD7/26]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC680745-500 BNC680745-100
BNC680745-500EA 4180 DKK
BNC680745-500 BNC680745-100
Anti-PTPRC Mouse Monoclonal Antibody (CF568) [clone: 2B11 PD7/26]
Antistoffer
Recognizes the CD45 leukocyte common antigen (LCA) family which is comprised of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portions of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals. Antibody to CD45 is useful in differential diagnosis of lymphoid tumors from non-hematopoietic undifferentiated neoplasms. A positive result with this MAb is highly indicativeMore Product Information