Anti-HSPD1 Mouse Monoclonal Antibody (CF594) [clone: GROEL/730]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC940730-500 BNC940730-100
BTIUBNC940730-500EA 4180 DKK
BTIUBNC940730-500 BTIUBNC940730-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF594) [clone: GROEL/730]
Antistoffer
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). Hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog. Clone GROEL730 reacts only with human and is useful in distinguishing human hsp60 from other mammalian and bacterial hsp60.

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: GROEL/730
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR