Anti-TG Mouse Monoclonal Antibody (CF568) [clone: 2H11 6E1]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC680724-500 BNC680724-100
BNC680724-500EA 4180 DKK
BNC680724-500 BNC680724-100
Anti-TG Mouse Monoclonal Antibody (CF568) [clone: 2H11 6E1]
Antistoffer
Thyroglobulin is a 660 kDa dimeric pre-protein with mutiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid originMore Product Information