Anti-MLANA Mouse Monoclonal Antibody [clone: M2-7C10 M2-9E3 A103]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNUB0700-500 BNUB0700-100
BNUB0700-500EA 3810 DKK
BNUB0700-500 BNUB0700-100
Anti-MLANA Mouse Monoclonal Antibody [clone: M2-7C10 M2-9E3 A103]
Antistoffer
This antibody recognizes a protein doublet of 20-22 kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum. This MAb cocktail labels melanomas and other tumors showing melanocytic differentiation. It is also aMore Product Information