Anti-MUC1 Mouse Monoclonal Antibody (Biotin) [clone: VU-2G7]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNCB0630-500 BNCB0630-100
BNCB0630-500EA 4180 DKK
BNCB0630-500 BNCB0630-100
Anti-MUC1 Mouse Monoclonal Antibody (Biotin) [clone: VU-2G7]
Antistoffer
MAb VU-2G7 reacts with MUC1, a large transmembrane glycoprotein expressed on the ductal surface of normal glandular epithelia. The dominant epitope of VU-2G7 has not been established with epitope fingerprinting. The extracellular domain of MUC1 largely consists of a highly conserved, O-glycosylated 20 amino acids tandem repeat which can occur 30-100 times per molecule depending on the length of the allele involved. In the vast majority of human carcinomas this protein is up regulated and poorly glycosylated and appears on the cell surface in a non-polarized fashion.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: VU-2G7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR