Anti-FUT4 Mouse Monoclonal Antibody [clone: FUT4/815 BRA-4F1]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNUB0562-500 BNUB0562-100
BNUB0562-500EA 3810 DKK
BNUB0562-500 BNUB0562-100
Anti-FUT4 Mouse Monoclonal Antibody [clone: FUT4/815 BRA-4F1]
Antistoffer
CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

  • Purified

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: FUT4/815 BRA-4F1
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR