Anti-PGR Mouse Monoclonal Antibody (CF488A) [clone: PR501]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC880501-500 BNC880501-100
BNC880501-500EA 4180 DKK
BNC880501-500 BNC880501-100
Anti-PGR Mouse Monoclonal Antibody (CF488A) [clone: PR501]
Antistoffer
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR501
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR