Anti-PGR Mouse Monoclonal Antibody (Biotin) [clone: PR500]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNCB0500-500 BNCB0500-100
BNCB0500-500EA 4180 DKK
BNCB0500-500 BNCB0500-100
Anti-PGR Mouse Monoclonal Antibody (Biotin) [clone: PR500]
Antistoffer
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR500
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human