Anti-Biotin Mouse Monoclonal Antibody (CF568) [clone: Hyb8]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC680400-500 BNC680400-100
BNC680400-500EA 4180 DKK
BNC680400-500 BNC680400-100
Anti-Biotin Mouse Monoclonal Antibody (CF568) [clone: Hyb8]
Antistoffer
This antibody recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Hyb8
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species