Anti-Bovine Serum Albumin Mouse Monoclonal Antibody (CF405S) [clone: ALB/398]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC040398-500 BNC040398-100
BNC040398-500EA 4180 DKK
BNC040398-500 BNC040398-100
Anti-Bovine Serum Albumin Mouse Monoclonal Antibody (CF405S) [clone: ALB/398]
Antistoffer
Reacts with a protein of ~66 kDa, identified as bovine serum albumin (BSA). It is a high affinity antibody and can be used for detection of traces of BSA. Bovine serum albumin (BSA) is an abundant plasma protein in cows that is important for maintaining osmotic pressure in blood plasma for proper distribution of body fluids between intravascular compartments and body tissues. BSA is a common buffer component for immunoglobulin type assays due to good solubility characteristics for water, Ca2 , Na , K , fatty acids, hormones and bilirubin. BSA makes up about half of the protein in plasma and represents the most stable and soluble protein in the plasma. It is a suitable reagent for laboratories developing immunoassays, mostly due to its availability, solubility and the numerous functional groups present for coupling. The BSA component contains several lysines that are capable of reacting with conjugation sites of linkers, making it applicable as a carrier protein for antigenic compounds.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: ALB/398
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR