Anti-NKX2-2 Mouse Monoclonal Antibody (Biotin) [clone: NX2/294]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNCB0294-500 BNCB0294-100
BNCB0294-500EA 4180 DKK
BNCB0294-500 BNCB0294-100
Anti-NKX2-2 Mouse Monoclonal Antibody (Biotin) [clone: NX2/294]
Antistoffer
Expression of NKX2.2 has been found in neuroendocrine tumors of the gut, making it a potential marker for the study of gastrointestinal neuroendocrine tumors. More recently, NKX2.2 protein was identified as a target of EWS-FLI-1, the fusion protein specific to Ewing sarcoma, and was shown to be differentially upregulated in Ewing sarcoma on the basis of array-based gene expression analysis. It acts as a valuable marker for Ewing sarcoma, with a sensitivity of 93% and a specificity of 89%, and aids in the differential diagnosis of small round cell tumors.
Type: Primary
Antigen: NKX2-2
Clonality: Monoclonal
Clone: NX2/294
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Chicken, Human, Mouse, Rat