Anti-Lung Specific Antigen Mouse Monoclonal Antibody (CF640R) [clone: LSA120]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC400120-500 BNC400120-100
BNC400120-500EA 4180 DKK
BNC400120-500 BNC400120-100
Anti-Lung Specific Antigen Mouse Monoclonal Antibody (CF640R) [clone: LSA120]
Antistoffer
LSA120 MAb is directed against a lung cell antigen. It shows a well-defined reactivity with cells of the lung lineage at all stages of maturation. Non-lung lineages are negative by immuno-histology. Although the exact identity of its antigen has yet to be determined, it has been shown to be distinct from MOC-52 and UM-A9 proteins. This MAb is useful in the identification of normal and neoplastic human lung cells.
Type: Primary
Antigen: Lung Specific Antigen
Clonality: Monoclonal
Clone: LSA120
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human